Order Entry
United States
ContactUsLinkComponent
Difco™ Beef Extract, Gibco™ (a part of Thermo Fisher Scientific)
Difco™ Beef Extract, Gibco™ (a part of Thermo Fisher Scientific)
Catalog # 76628-838
CAS Number:  
Difco™ Beef Extract, Gibco™ (a part of Thermo Fisher Scientific)
Catalog # 76628-838
Supplier Number:  212610
CAS Number:  

Specifications

  • Description:
    Difco™ Beef extract
  • Size:
    500 g
  • Cat. No.:
    76628-838
  • Source:
    Animal origin: Bovine

Specifications

About this item

Difco Beef extract is an nutritional supplement provides a mixture of peptides, individual amino acids, nucleotides, organic acids, minerals, and vitamins.

  • Derived from infusion of beef
  • Provides a rich source of nutrients
  • Available as a paste Intended to replace aqueous infusion of meat in microbiological culture media

Provided as a paste, customers have used Difco Beef extract in a variety of microbial fermentation cell lines, including Escherichia coli, Bacillus subtillis, Pasteurella multocida, and Streptococcus species to produce numerous animal health vaccines.