Motic AE2000 Inverted Microscope LED
About this item
The AE2000 is a revolutionary design that is packed with features and sets a new benchmark in affordable inverted microscopy. Motic’s AE2000 inverted microscope is designed for routine live cell inspection with an easy and flexible set up, ideal for university, routine-clinical, and lab environments.
- Head Siedentopf type 45° inclined
- Side facing quadruple nosepiece
- CCIS plan achromat objective PL PH4X, PL PH 10X (basic package)
- Optional CCIS plan achromat objective with PL PH 20X
- ELWD N.A. 0.30 condenser
- Phase slider Ph0, BDF, Ph1
- Centering telescope
- Plain stage with metal and glass stage inserts
- 3 W LED illumination with intensity control
Binocular inverted live cell microscope for routine microbiology. The pre-defined optical setup with LWD CCIS© Phase objectives is ready for the inspection of diverse native samples in cell culture flasks. Lab technicians benefit from the convincing phase contrast concept, while the Fixed-Köhler illumination with interchangeable light source (HAL/LED) and auto ON/OFF function enables a fast throughput of samples.
Ordering information: Contact your Avantor representative to learn more about this and other motic solutions. All motic products come with a one-year electrical and five-year mechanical warranty.
4 Options Available Below
Product Details & Documents
The AE2000 is a revolutionary design that is packed with features and sets a new benchmark in affordable inverted microscopy. Motic’s AE2000 inverted microscope is designed for routine live cell inspection with an easy and flexible set up, ideal for university, routine-clinical, and lab environments.
- Head Siedentopf type 45° inclined
- Side facing quadruple nosepiece
- CCIS plan achromat objective PL PH4X, PL PH 10X (basic package)
- Optional CCIS plan achromat objective with PL PH 20X
- ELWD N.A. 0.30 condenser
- Phase slider Ph0, BDF, Ph1
- Centering telescope
- Plain stage with metal and glass stage inserts
- 3 W LED illumination with intensity control
Binocular inverted live cell microscope for routine microbiology. The pre-defined optical setup with LWD CCIS© Phase objectives is ready for the inspection of diverse native samples in cell culture flasks. Lab technicians benefit from the convincing phase contrast concept, while the Fixed-Köhler illumination with interchangeable light source (HAL/LED) and auto ON/OFF function enables a fast throughput of samples.
Ordering information: Contact your Avantor representative to learn more about this and other motic solutions. All motic products come with a one-year electrical and five-year mechanical warranty.
Documents
Product Family Options
Product Information
- AvailabilityPrice
- 76595-444AE2000 binocular inverted microscope LED, basic packageSupplier Number: 1100103800222
- 76595-450AE2000 binocular inverted microscope LED, with PL PH 20XSupplier Number: 1100103800268
- 76595-446AE2000 trinocular inverted microscope LED, basic packageSupplier Number: 1100103800266
- 76595-448AE2000 trinocular inverted microscope LED, with PL PH 20XSupplier Number: 1100103800267
Accessories
Product Information
- Catalog #AvailabilityPrice
- Glass stage insert
Product Information
- Catalog #AvailabilityPrice
- CCIS Plan Achromat Phase PL Ph4x, N.A. 0.10 / W.D. 12.6, Ph0CCIS Plan Achromat Phase PL Ph4x, N.A. 0.10 / W.D. 12.6, Ph089168-402Log in for availabilityEach - $188.51
Product Information
- Catalog #AvailabilityPrice
- CCIS Plan Achromat LWD PL 20X N.A.0.3 / W.D. 4.7
Product Information
- Catalog #AvailabilityPrice
- Widefield Eyepiece N-WF 15x/16mm
Product Information
- Catalog #AvailabilityPrice
- Phase Stop Ph0 (PL Ph4x)
Product Information
- Catalog #AvailabilityPrice
- Hemacytometer holder
Product Information
- Catalog #AvailabilityPrice
- 0.5x C-Mount Camera Adapter (for 0.5" Chip Sensors)0.5x C-Mount Camera Adapter (for 0.5" Chip Sensors)10767-528Log in for availabilityEach - $185.54
Product Information
- Catalog #AvailabilityPrice
- CCIS Plan Achromat LWD PL 40X N.A. 0.5 / W.D. 3
Product Information
- Catalog #AvailabilityPrice
- Phase slider, centerable, BF, centerable
Product Information
- Catalog #AvailabilityPrice
- 0.65x C-Mount Camera Adapter (for 0.66" Chip Sensors)0.65x C-Mount Camera Adapter (for 0.66" Chip Sensors)10767-530Log in for availabilityEach - $202.45
Product Information
- Catalog #AvailabilityPrice
- 1x C-Mount Camera Adapter (no Lens)
Product Information
- Catalog #AvailabilityPrice
- Phase Stop Ph1 (PL Ph 10x, PL Ph 20x)
Product Information
- Catalog #AvailabilityPrice
- CCIS Plan Achromat PL 10X N.A. 0.25 / W.D. 16.8