Motic BA410E LED Sextuple Compound Microscope
Supplier: Motic
Certificates
About this item
The BA410E is Motic's flagship model, and it displays the highest level of optical performance. It also allows a profound diagnosis for those in pathology, haematology and cytology who require the best color fidelity. Increased numerical apertures provide an expanded resolution power for a more detailed specimen evaluation.
- Head Siedentopf type 30° inclined, 360° rotating [trinocular: (light split = ²⁰/₈₀)]
- Reversed sextuple nosepiece
- Coaxial coarse and fine focusing adjustment
- Built in low position rackless stage with right hand control
- Achromat swing-out condenser N.A. 0.90 with iris diaphragm
Binocular or trinocular research microscope for diverse questions from life sciences, but also from material sciences. The standard EC-H plan achromat objectives can be replaced by fluorite and apochromatic lenses for maximum optical performance. Demanding users benefit from the system’s flexibility of the BA410E construction kit, ready to configure a personal microscope for individual requirements.
Ordering information: Contact your Avantor representative to learn more about this and other motic solutions. All motic products come with a one-year electrical and five-year mechanical warranty.
Related Information
Contrast Methods, Darkfield with Slider Solution for BA Series Contrast Methods, Polarizing System How to Adjust the Microscope Illumination How to Adjust the Tension of the Focusing System (Ring). E.g. AE Series How to Avoid Repairs in your Microscope
Videos
More
Motic Microscopy Solutions Motic Microscopy Solutions.pdf
Product Literature
Frequently Bought Together