PCR Plates, 96-Well, Semi-Skirted, ABI Style with Upstand, Azenta Life Sciences
Supplier: AZENTA US, INC
Certificates
About this item
These plates are designed for use on ABI thermal cyclers. 0.2 ml wells, polypropylene, cut corner H12; working volume: <200 µl, total well capacity: 300 µl.
- Subject to a strict QC procedure which includes a visual check and leak testing of every tube
- Free from DNase, RNase, human genomic DNA and endotoxin/pyrogen
- Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
- Standard 0.2 ml (200 µl) working volume, 0.3 ml (300 µl) total well capacity when used with sealing options
- Maximum thermal conductivity for efficient heat transfer and precise thermal cycling
- Raised well rims prevent cross contamination and facilitate effective sealing for reduced evaporation
- Alphanumeric grid reference to aid well and sample identification
- Recommended for low volume PCR
- Compatible with standard multichannel pipettes
We recommend this semi-skirted plate for use with ABI® thermal cyclers and sequencers. It can be used directly with ABI® instruments with no adapters and no re-calibration of the instruments necessary. The only case where this is not true is with the ABI Fast Block thermal cyclers, in which case using the FrameStar FastPlate is recommended instead.
All of these PCR plates are molded from virgin polypropylene under ISO certified cleanroom conditions in a UK-based production facility and as such comply with the same stringent requirements as the FrameStar® range.
Ordering information: Available barcoded upon request.