Order Entry
United States
ContactUsLinkComponent
 
FrameStar® PCR Plates, 96-Well, Fully Skirted, Flat Optical Bottom, Low Profile, Azenta Life Sciences
Catalog # 76629-050
Supplier:  AZENTA US, INC
undefined
FrameStar® PCR Plates, 96-Well, Fully Skirted, Flat Optical Bottom, Low Profile, Azenta Life Sciences
Catalog # 76629-050
Supplier:  AZENTA US, INC
Supplier Number:  4TI-0970

Specifications

  • Frame color:
    Clear
  • No. of wells:
    96
  • Profile:
    Low
  • Skirt:
    Fully skirted
  • Well Color:
    Black
  • Well volume:
    0.1 ml
  • Description:
    PCR plate, PP wells, PC frame, cut corner H1, optical bottom
  • Environmentally Preferable:
  • Cat. No.:
    76629-050

Specifications

About this item

Flat optically clear bottom, low profile, 0.1 ml polypropylene wells, polycarbonate frame, cut corner H1; working volume: <100 µl, total well capacity: 200 µl. Ideal for use with robotic systems. Compatible with standard multichannel pipettes.

  • Rigid polycarbonate frame for added mechanical stability
  • <0.1 ml (100 µl) working volume, 0.18 ml (180 µl) total well capacity
  • Improved seal integrity due to thermal stability of frame
  • Alphanumeric grid reference to aid well and sample identification
  • Recommended for low volume PCR
  • Suitable for microscopy and small volume sample storage.

Free from DNase, RNase, human genomic DNA, and endotoxin or pyrogen stackable. Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results. Significantly reduced thermal expansion and sample evaporation for Improved consistency in PCR results.

The FrameStar 96 well skirted optical bottom PCR plates are unique in the market, being suitable for use in both microscopy and PCR. Optical bottom plates are ideal for applications requiring single cell sorting followed by molecular biology techniques such as (q)PCR and sequencing. The flat bottoms enable excellent stack ability, making these plates well suited for small sample volume storage such as compound libraries, with no risk of damaging the seal of the plate below. Additionally, the small well volume enables excellent sample recovery.

The low profile wells of this plate are shorter than ‘standard’ profile wells, which decreases the ‘dead space’ between the heated lid of the thermal cycler and the reagents in the well. This reduces condensation forming on the side wall of the tubes, moderating changes in PCR volume and increasing the efficiency of the reaction. This is especially recommended for reaction volumes below 20 µl.

The rigid polycarbonate skirt of this design eliminates warping and distortion during PCR and makes it ideal for use with robotic systems.

Ordering information: Available barcoded upon request.