Low profile, 0.1 ml polypropylene wells, polycarbonate frame, cut corner H1; working volume: <100 µl, total well capacity: 200 µl.
- Rigid polycarbonate frame for added mechanical stability
- <0.1 ml (100 µl) working volume, 0.2 ml (200 µl) total well capacity
- Improved seal integrity due to thermal stability of frame
- Alphanumeric grid reference to aid well and sample identification
- Greatest compatibility with different thermal cyclers
- Recommended for low volume PCR
Free from DNase, RNase, human genomic DNA, and endotoxin or pyrogen stackable. Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results. Significantly reduced thermal expansion and sample evaporation for improved consistency in PCR results. Compatible with standard multichannel pipettes.
This universal non-skirted plate is designed for use on all major thermal cyclers.
The low profile wells of this plate are shorter than ‘standard’ profile wells, which decrease the ‘dead space’ between the heated lid of the thermal cycler and the sample in the well. This eliminates condensation forming on the side wall of the tubes, preventing reduction in PCR volume and increasing the efficiency of the reaction.
The rigid polycarbonate skirt of this design eliminates warping and distortion during PCR, meaning seal integrity is not compromised and less of your sample is lost through evaporation. This is especially recommended for reaction volumes below 20 µl.