PCR Plates, 96-Well, Semi-Skirted, Azenta Life Sciences
Supplier: AZENTA US, INC
Certificates
About this item
Universal 96-well skirted plate designed for use on standard thermal cyclers. 0.2 ml wells, polypropylene, cut corner A12; working volume: <200 µl, total well capacity: 300 µl.
- Subject to a strict QC procedure which includes a visual check and leak testing of every tube
- Free from DNase, RNase, human genomic DNA and endotoxin/pyrogen
- Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
- High profile 0.2 ml (200 µl) working volume, 0.3 ml (300 µl) total well capacity when used with sealing options
- Maximum thermal conductivity for efficient heat transfer and precise thermal cycling
- Raised well rims prevent cross contamination and facilitate effective sealing for reduced evaporation
- Alphanumeric grid reference to aid well and sample identification
- Recommended for low volume PCR
- Compatible with universal standard block thermal cyclers, sequencers and standard multichannel pipettes
All of these PCR plates are molded from virgin polypropylene under ISO certified cleanroom conditions in a UK-based production facility, and as such comply with the same stringent requirements as the FrameStar range.
Frequently Bought Together