Order Entry
United States
ContactUsLinkComponent
 
Prostate Biopsy Fixative with Eosin, StatLab
Catalog # 76548-188
Supplier:  StatLab
CAS Number:  
undefined
Prostate Biopsy Fixative with Eosin, StatLab
Catalog # 76548-188
Supplier:  StatLab
Supplier Number:  PF0507B500
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Prostate Biopsy Fixative with Eosin
  • Size:
    20 ml (500 per case)
  • Cat. No.:
    76548-188

Specifications

About this item

20mL StatClick™ Vials filled with a special formulation for prostate biopsies. Contains formalin, alcohol, and has a slight Eosin tint to ensure easy identification of biopsies after processing and embedding. StatClick patented vials produce an audible “clicking” sound when the leak-proof seal has been properly engaged, assuring accurate closure and maintaining specimen integrity during transport.

Packaging: Manufactured in the USA