Urofix Fixative for Prostate Biopsies, StatLab
Supplier: StatLab
Certificates
About this item
An excellent fixative for prostate biopsies. Preserves nuclear detail extremely well, and penetrates tissue quicker, thereby shortening fixation times.
Packaging: Manufactured in the USA
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.