Order Entry
United States
ContactUsLinkComponent
TissueGuard™ Specimen Processing Gel, StatLab
TissueGuard™ Specimen Processing Gel, StatLab
Catalog # 76548-294
Supplier:  StatLab
CAS Number:  
undefined
TissueGuard™ Specimen Processing Gel, StatLab
Catalog # 76548-294
Supplier:  StatLab
Supplier Number:  TG12
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    TissueGuard™ Specimen Processing Gel
  • Size:
    12 per box
  • Cat. No.:
    76548-294
  • Volume:
    10 ml

Specifications

About this item

tabilize fragile specimens during processing by preparing with TissueGuard processing gel. Tissue Guard encapsulates and retains specimens during histological processing while allow chemical reagents to penetrate the specimen. Appropriate for cytology and histology applications, TissueGuard is an alternate to other agarose stabilizers.

  • 12 tubes per kit
  • 10 mL per tube

Cytology Protect fine needle aspirates, urine specimens, non-gyn specimens, and any other specimens utilizing cell block preparation. Product can be applied in either centrifuge tube, or in conjunction with BioMesh tissue cassettes, or using tissue paper. After application, specimen is ready for processing within minutes, or sooner if aided with a cooling block. Histology Protect samples including needle biopsies, nerves, small arteries, aggregates, lymph nodes and the smallest of tissue fragments. Product can be applied to samples in conjunction with BioMesh cassettes, or using filter paper.

Packaging: Manufactured in the USA