Order Entry
United States
Orders LinkContactUsLinkComponent
Tissue Marking Dyes, StatLab

Tissue Marking Dyes, StatLab

Supplier:   StatLab
Tissue Marking Dyes, StatLab
Supplier:  StatLab
Certificates
Tissue Marking Dyes, StatLab
Tissue Marking Dyes, StatLab

8 Options Available Below

Product Details & Documents

Permanent tissue marking dyes. Seven colors formulated to define margins without bleeding or fading. Durable, bright dyes hold permanently through processing and staining procedures. Unique lids may either be completely removed or conveniently flipped open to prevent drying.

Set includes one tissue marking dye rack and a set of 2 oz dyes in each of 7 colors.

Packaging: Manufactured in the USA

Product Family Options

Product Information

  • Color
    Size
    Availability
    Price
  • 76548-240
    Margin Marker
    Margin Marker
    Black
    2 oz
    Log in for availability
    Log in for availability
    Each (2oz)
    Each (2oz) - $43.12
    $43.12
    Each (2oz)
       $43.12

    Supplier Number: SL662BK-2
  • 76548-242
    Margin Marker
    Margin Marker
    Blue
    2 oz
    Log in for availability
    Log in for availability
    Each (2oz)
    Each (2oz) - $42.16
    $42.16
    Each (2oz)
       $42.16

    Supplier Number: SL662BL-2
  • 76548-244
    Margin Marker
    Margin Marker
    Green
    2 oz
    Log in for availability
    Log in for availability
    Each (2oz)
    Each (2oz) - $42.64
    $42.64
    Each (2oz)
       $42.64

    Supplier Number: SL662GR-2
  • 76548-246
    Margin Marker
    Margin Marker
    Orange
    2 oz
    Log in for availability
    Log in for availability
    Each (2oz)
    Each (2oz) - $41.22
    $41.22
    Each (2oz)
       $41.22

    Supplier Number: SL662OR-2
  • 76548-248
    Margin Marker
    Margin Marker
    Red
    2 oz
    Log in for availability
    Log in for availability
    Each (2oz)
    Each (2oz) - $42.38
    $42.38
    Each (2oz)
       $42.38

    Supplier Number: SL662RD-2
  • 76548-250
    Margin Marker
    Margin Marker
    Violet
    2 oz
    Log in for availability
    Log in for availability
    Each (2oz)
    Each (2oz) - $42.10
    $42.10
    Each (2oz)
       $42.10

    Supplier Number: SL662VT-2
  • 76548-252
    Margin Marker
    Margin Marker
    Yellow
    2 oz
    Log in for availability
    Log in for availability
    Each (2oz)
    Each (2oz) - $41.88
    $41.88
    Each (2oz)
       $41.88

    Supplier Number: SL662YL-2
  • 76548-254
    Tissue Marking Dye Set
    Tissue Marking Dye Set
    Assorted colours
    1 case
    Log in for availability
    Log in for availability
    Case (2oz)
    Case (2oz) - $332.46
    $332.46
    Case (2oz)
       $332.46

    Supplier Number: SL669
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.