Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:1% Focus™ Acid Alcohol
- Size:1 Gallon
- Cat. No.:76540-468
Specifications
About this item
Contains hydrochloric acid in 70% reagent grade alcohol. Used to remove excess stain and to produce better defined nuclei.
Available in 0.5% and 1.0% formulations.