- Description:Coverslipping Tape, 70 m
- Packed:5 per case
- Cat. No.:76548-146
An exclusive resin coated, tri-acetate film that eliminates the need for glass coverslips.
- Manufactured using a proprietary micro-diffusion adhesive, which helps to facilitate optimal adhesive dispersion
- The film exhibits excellent transparency and optical performance, without the long-term formation of bubbles or edge lifting
- Each roll is 70 meters in length, 5 rolls per case
Part of the StatLab family of coverslipping tape. Manufactured in Japan.
Millennia coverslipping tape is validated by Hologic® for use with Hologic’s ThinPrep® Imaging Systems. Please contact Hologic with any questions.