StatCut™ Microtome Blades, StatLab Medical Products
Supplier: StatLab
Certificates
About this item
Extra-sharp blades for precise cutting at the microtome.
- Suitable for both soft and hard tissue
- Well balanced in durability and sharpness
- Coated for non-stick sections and effortless ribboning
Blades can accommodate high volume laboratories. Manufactured in Japan.
Packaging: Blades are packaged 50 per pack.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.