Sodium dihydrogen phosphate monohydrate USP endotoxin tested GMP Manufactured Product
Catalog # JT4954-60
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Size:50 kg
- Cat. No.:JT4954-60
Specifications
Specification Test Results
- USP - Assay (anhydrous basis) :98.0 - 103.0 %
- USP - Aluminum, Calcium, and Related Elements:Passes Test
- USP - Arsenic (As):≤ 3 ppm
- USP - Chloride (Cl) :≤ 0.014 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Insoluble Substances:≤ 0.2 %
- USP - pH (1 in 20) :4.1 - 4.5
- USP - Sulfate (SO₄) :≤ 0.15 %
- USP - Water (H₂O):10.0 - 15.0 %
- Endotoxin Concentration (EU/g:≤ 2.5
- Total Aerobic Microbial Coun:≤ 100 CFU/g
- Total Yeast and Mold Count:≤ 50 CFU/g
Specification Test Results
About this item
:
:137.99 g/mol
Merck Index:13,08734
Melting Pt:100 °C
Density:2.04 g/cm³
Storage Temperature:Ambient
EINECS:231-449-2
REACH:01-2119489796-13
CAS Number:10049-21-5
Formula:NaH₂PO₄·H₂O
MDL Number:MFCD00149208