Specifications
- Orbit:3 mm
- Power:200 W
- Description:Microtube shaker
- Temperature Accuracy:0.5 °C
- Overall Dimensions:23.5W×20D×12H cm
- Temperature Uniformity:0.5 °C
- Heat-up Time:5.5 °C/min
- Cooling Speed:5 °C/min (100 °C to room temperature); 0.5 °C/min (below room temperature)
- Mixing speed range:200 - 1500 rpm
- Programs:Up to 6 steps per 9 programs
- Temperature Setting:0.1...100 °C
- Weight:7.3 kg
- Cat. No.:76545-350
- Voltage:115 V
Specifications
About this item
Compact benchtop shaking incubator ideal for molecular biology applications. Uniformly heat, cool and shake samples within microtubes or microplates on demand. Requires no tools for block exchange via magnetic connection technology. Programmable up to six steps per nine programs. Can be used in a cold room environment.
- Quick interchange of blocks with magnetic adhesion technology
- Calibration mode ensures accurate temperatures within ±0.5 °C after every block exchange
- Flexible block options and versatile tube/plate compatibility
- Safe for cold room use
- Stable self-recognizing block
- Constant mixing, wide speed range, programmable
The Axygen Microtube Shaker provides researchers with a benchtop shaking incubator that allows uniform heating, cooling and mixing of precious life science samples. The compact footprint incorporates an intuitive control panel which allows the operator to easily program the device and view the live temperature, time and mixing speed settings throughout the experiment.
Nine interchangeable aluminum blocks accommodate PCR plates and a variety of common life science vessels. No additional tools are required for block exchanges. The blocks utilize magnetic adhesion technology and are self-recognized once exchanged, simply mount and exchange blocks in seconds. Mixing speeds vary by volume and range from 200-1500 rpm. The combination of heating, cooling and mixing with a versatile block selection makes the Axygen Microtube Shaker ideal for many life science laboratories in molecular biology, biochemistry, and clinical chemistry area. Applications include but are not limited to gene synthesis, gene purification and denaturation, enzymatic reactions, and bacterial growth assays.