Order Entry
United States
ContactUsLinkComponent
 
EndoTrap® HD 1/1, Biovendor
Catalog # 103853-666
Supplier:  BioVendor
CAS Number:  
EndoTrap® HD 1/1, Biovendor
Catalog # 103853-666
Supplier:  BioVendor
Supplier Number:  LET0009
CAS Number:  

Specifications

  • Cat. No.:
    103853-666

Specifications

About this item

EndoTrap® HD removes endotoxins (lipopolysaccharides/LPS) from aqueous solutions of a wide range of samples such as proteins, antibodies, vaccines, nucleic acids, buffers and cell extracts with excellent sample recovery rate.

The EndoTrap® HD resin is especially designed to meet the needs of pharma and biotech industry for the efficient removal of endotoxins in biomanufacturing processes as well as any application that requires highest endotoxin removal rates.

EndoTrap® HD fulfils all requirements for industrial applications, and it can easily be integrated in/at early or late-stage purification processes. Moreover, the High-Definition Matrix is also available for small-scale purification as pre-filled columns.

Endotoxin removal with EndoTrap® HD works effectively in the pH range of 4-9 and NaCl concentrations of 50-600 mM. Buffers such as HEPES, TRIS, MOPS, MES, and PIPES are recommended. Citrate buffers and chelators of divalent cations (like EDTA) have to be avoided

  • Best-in-class endotoxin binding capacity of > 5,000,000 EU/ml
  • Removes more than 99.99% of LPS
  • Excellent sample recovery rate >95%
  • Non-toxic bacteriophage-derived ligand, no cytotoxic effects
  • Re-use more than 10 times
  • pH sample range: 4-10
  • suitable for high salt conditions
  • suitable for e.g. TRIS, HEPES, MOPS or similar buffered samples
  • PBS buffer possible when enriched freshly with 0.1 mM calcium (magnesium) directly before use
  • EndoTrap® Leakage ELISA for ligand leakage monitoring
  • Available as resin (buffers on request).
Delivery: 1 x 1 ml column, 25 mL 5x Equilibration Buffer, 25 mL 5x Regeneration Buffer, 25 mL 5x Storage Buffer