Order Entry
United States
ContactUsLinkComponent
Buck/Boost Step-Up Prewired Transformer, Single Phase, Larson Electronics LLC
Buck/Boost Step-Up Prewired Transformer, Single Phase, Larson Electronics LLC
Catalog # MSPP-MPDMTBBT2
Irregular Voltage
Buck/Boost Step-Up Prewired Transformer, Single Phase, Larson Electronics LLC
Catalog # MSPP-MPDMTBBT2
Supplier Number:  MPD-MT-BBT
Irregular Voltage

Specifications

  • Output:
    240 V
  • Description:
    Tranformer single-phase step-up prewired
  • Environmentally Preferable:
  • Cat. No.:
    MSPP-MPDMTBBT2
  • L×W×H:
    29.8×16.5×17.8 cm (11.75×6.5×7")
  • Electrical:
    208 V, 50/60 Hz
  • Weight:
    16.3 kg (36 lbs.)

Specifications

About this item

The MPD-MT-BBT-240V-208V-20A-2X3FT-L6.20 Single phase step-up buck and boost prewired transformer from Larson Electronics is powerful, reliable, and designed with the environment in mind. This transformer is designed to increase 240 to 208 V and is needed when the line-voltage is lower than the required voltage for equipment.

  • Suitable for both indoor and outdoor applications, the MPD-MT-BBT-240V-208V-20A-2X3FT-L6.20 provides:
  • increased reliability
  • higher efficiency
  • protection against critical equipment failures
  • extra level of protection by isolating the power source from the connected device
  • lower operating costs
  • lower heat emissions
  • lower cost of ownership

The MPD-MT-BBT-240V-208V-20A-2X3FT-L6.20 is a prewired transformer for customer installation and is equipped with 3’ of 12/3 SOOW cable on each side connected to the transformer with a 20 amp L6- 20P twist lock plug on the primary side and a 20 amp L6-20C twist lock receptacle on the secondary side.

Buck and boost transformers are designed to maximize the performance and life of electrical equipment. The MPD-MT-BBT-208V-230V-20A-2X3FT-6.20 is encapsulated in silica sand and resin, and cased in a NEMA 3R steel enclosure. NEMA 4, NEMA 4X, and NEMA 12 enclosures are available upon request.