ContactUsLinkComponent
Popular Science by Celestron Outland X 10×32 mm Roof Binocular with Smartphone Adapter and Bluetooth Remote
Popular Science by Celestron Outland X 10×32 mm Roof Binocular with Smartphone Adapter and Bluetooth Remote
Catalog # 470347-580
Supplier:  Avantor
undefined
Popular Science by Celestron Outland X 10×32 mm Roof Binocular with Smartphone Adapter and Bluetooth Remote
Catalog # 470347-580
Supplier:  Avantor
Specifications
  • Magnification:
    10×
  • Description:
    Outland X 10×32 mm roof binocular
  • Cat. No.:
    470347-580
Specifications
About this item
Observe wildlife at a distance and share captured photos and videos instantly.
  • Strong 10× magnification to bring distant subjects up close
  • Smartphone adapter and Bluetooth remote included to capture still images or video through the binocular
  • Eyeglass friendly with generous eye relief and sturdy, twist-up eyecups with multiple stops
  • Tripod adaptable
BaK-4 prisms and multi-coated optics provide improved contrast and resolution by increasing light transmission through the entire optical path. Rubber armored for improved ergonomics and protection from everyday use in the field.