Light Emitting Diodes
Supplier: United Scientific Supplies
Certificates
About this item
Quality LEDs for electrical experiments.
- 2 pin connectors
- Fits all standard bred boards
- Available in two sizes
These LEDs fit perfectly into a Bred board for investigating electrical circuits. By their nature, these LEDs will only work correctly in DC circuits with the proper electrical orientation. Each LED has a Cathode (– shorter side) and Anode (+ longer side).
Frequently Bought Together