Specifications
- Method / Format:Magnetic beads
- Prep size:Scalable
- Refine target molecule:Total DNA
- Sample type:Biofluids (serum, plasma, saliva)
- Target molecule:DNA
- Description:Mag-Bind® blood and tissue DNA HDQ prefilled 96 kit
- Size:4×96 preparations
- Cat. No.:76492-582
Specifications
About this item
Automation-ready, prefilled 96-well plates for quick and convenient DNA extraction from blood, saliva, cultured cells, and swabs for magnetic processors.
- Ease of Use – Reduction of hands-on time with no reagent preparation (for bind, wash, and elute steps)
- Convenience – Inclusion of tip combs and other required plasticware in the kit for user convenience
- Extraction Accuracy – Reduction of user-associated errors
- Safe – No organic solvents
- High-Quality – Purified DNA suitable for downstream applications such as next-generation sequencing, qPCR, PCR, and microarray
The Mag-Bind® Blood and Tissue DNA HDQ Prefilled 96 Kit includes prefilled plates in 96-well standard automation format. The configuration of this kit works seamlessly on magnetic processors such as the Thermo Fisher Scientific KingFisher® Flex, QIAGEN BioSprint®, and Applied Biosystems MagMAX® Express-96. Included in this kit are all plastic accessories, including tip combs, that are needed for an extraction run post-lysis. This kit offers a versatile method for isolating high-quality DNA from a wide variety of samples, including fresh or frozen animal cultured cells and tissues, buccal swabs, mouse tail snips, dried blood spots, up to 250 μl whole blood, and up to 250 μl saliva. With no reagent set-up and the quick magnetic response of the Mag-Bind® Particles PF-HDQ, hands-on time and the overall processing time are significantly reduced. The purification procedure provides high-quality DNA suitable for direct use in most downstream applications including amplification, next generation sequencing (NGS), and enzymatic reactions.