Order Entry
United States
ContactUsLinkComponent
Betamethasone Sodium Phosphate, Reference Standard, USP
Betamethasone Sodium Phosphate, Reference Standard, USP
Catalog # 101094-398
Supplier:  USP
CAS Number:  
Betamethasone Sodium Phosphate, Reference Standard, USP
Catalog # 101094-398
Supplier:  USP
Supplier Number:  1068004
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Vial
  • CAS No.:
    151-73-5
  • Description:
    Betamethasone Sodium Phosphate, Reference Standard
  • Size:
    500 mg
  • Environmentally Preferable:
  • UN No.:
    2811
  • Formula:
    C₂₂H₃₀FNa₂O₈P
  • Description:
    Betamethasone Sodium Phosphate
  • Cat. No.:
    101094-398
  • Supplier no.:
    1068004

Specifications

About this item

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard