Sol-Care® Safety Winged Blood Collection Needles, Sol M
Supplier: SOL-MILLENNIUM MEDICAL
Certificates
About this item
This safety-engineered device has a sliding safety shield that clicks when activated, alerting you that the needle is safely locked.
- Transparent design that allows visualization of the blood flashback and flow
- Single-handed activation option that provides more flexibility to care for the patient and venipuncture site
- Sliding safety shield pushes over the needle and locks with an audible click, protecting against needlestick injury
Available with or without pre-attached holder.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.