ContactUsLinkComponent
Adjustable Boss Head Clamp
Adjustable Boss Head Clamp
Catalog # 470343-240
Supplier:  GSC International
undefined
Adjustable Boss Head Clamp
Catalog # 470343-240
Supplier:  GSC International
Supplier Number:  4-ACH40-10
Specifications
  • Description:
    Boss head clamp
  • Cat. No.:
    470343-240
Specifications
About this item
These boss head clamp holders are adjustable up to 16 mm.
  • Accepts rods up to 5/8" in diameter
  • Made with cast-iron body and nickel-plated thumb-screws