Order Entry
United States
ContactUsLinkComponent
 
Louvered Wall Panel, Akro-Mils
  62661-490
 :  Akro-Mils
Louvered Wall Panel, Akro-Mils
  62661-490
 :  Akro-Mils
 :  30636

 

  • Width:
    35¾"
  • Color:
    Gray
  • Description:
    Wall-Mounted Bin Panel Hanging System
  • Height:
    19"
  • Cat. No.:
    62661-490
  • Supplier no.:
    30-636

 

 

Wall-mounted louvered panels keep parts up off the ground and free up floor space.

  • Formed from 16-gauge cold-rolled steel for strength and durability
  • Durable powder-coated finish stands up to heavy-duty service
  • Single-sided

For use with AkroBins or InSight Bins (sold separately).

AkroBins