ContactUsLinkComponent
Medical Chemical Corporation Carbol Fuchsin, Ziehl-Neelsen, for AFB Stain for Mycobacteria, Hardy Diagnostics
Medical Chemical Corporation Carbol Fuchsin, Ziehl-Neelsen, for AFB Stain for Mycobacteria, Hardy Diagnostics
  76591-146
 :  
undefined
Medical Chemical Corporation Carbol Fuchsin, Ziehl-Neelsen, for AFB Stain for Mycobacteria, Hardy Diagnostics
  76591-146
 :  484A8
 :  
  • Description:
    Carbol fuchsin, Ziehl-Neelsen, for AFB stain for mycobacteria
  • Size:
    8 oz.
  • Cat. No.:
    76591-146
An aqueous solution of basic fuchsin, phenol and reagent alcohol. ‘Hot method’, requires heating of the slide.
 : Ship ground.