ContactUsLinkComponent
Acid Fast Stain Kit with Brilliant Green Counterstain
Acid Fast Stain Kit with Brilliant Green Counterstain
Catalog # 76591-144
Supplier:  Hardy Diagnostics
CAS Number:  
undefined
Acid Fast Stain Kit with Brilliant Green Counterstain
Catalog # 76591-144
Supplier:  Hardy Diagnostics
Supplier Number:  483KB
CAS Number:  
Specifications
  • Description:
    AFB brilliant green stain kit
  • Cat. No.:
    76591-144
Specifications
About this item
The acid fast stain kit is used as a differential method to detect the difference between those organisms which decolorize with treatment of acid alcohol, and those that do not, such as the mycobacteria. ‘Cold Stain’ method.
Delivery: Ship ground.