Medical Chemical Corporation Acridine Orange Stain, 8 oz.
Catalog # 76591-136
Supplier: Hardy Diagnostics
CAS Number:
Specifications
- Description:Acridine orange stain
- Size:8 oz.
- Cat. No.:76591-136
- Application:Clinical
Specifications
About this item
Acridine Orange (fluorescent stain for bacteria in body fluids) 0.025% weight by volume. An aqueous solution of acridine orange.
Delivery: Ship ground.