Order Entry
United States
Orders LinkContactUsLinkComponent
2-Acetylpyrazine

2-Acetylpyrazine

Irritant
Supplier:   APOLLO SCIENTIFIC
2-Acetylpyrazine
Supplier:  APOLLO SCIENTIFIC
Certificates
2-Acetylpyrazine

1 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Pyrazin-2-ylethan-1-one, Acetylpyrazine
  • CAS Number:22047-25-2
  • Formula:C₆H₆N₂O
  • MDL Number:MFCD00006134
  • Molecular Weight:122.13 g/mol
  • Boiling Pt:78…79 °C/8mmHg
  • Melting Pt:76…78 °C
  • Storage Temperature:Ambient
  • EINECS:244-753-5
Product Family Options

Product Information

  • Size
    Pack type
    Availability
    Price
  • 5 g
    Bottle
    Discontinued
    Supplier Number: -
  • 25 g
    Bottle
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $21.34
    $21.34
    Each(1/Itm/Bx)
       $21.34

    Supplier Number: OR5593-25G
  • 100 g
    Bottle
    Discontinued
    Supplier Number: -

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.