Dexamethasone 96%
Supplier: Thermo Scientific Chemicals
About this item
Synonyms: Dexamethasonum, 9-Fluor-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadien-3,20-dion
- CAS Number:50-02-2
- Formula:C₂₂H₂₉FO₅
- MDL Number:MFCD00064136
- Molecular Weight:392.47 g/mol
- Melting Pt:255…261 °C (dec.)
- Storage Temperature:Ambient
- EINECS:200-003-9
3 Options Available Below
Product Details & Documents
Synonyms: Dexamethasonum, 9-Fluor-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadien-3,20-dion
- CAS Number:50-02-2
- Formula:C₂₂H₂₉FO₅
- MDL Number:MFCD00064136
- Molecular Weight:392.47 g/mol
- Melting Pt:255…261 °C (dec.)
- Storage Temperature:Ambient
- EINECS:200-003-9
Specifications for Product Family
Specification Test Results
- Appearance:White to almost white Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (500 mg, 105°C)
- UV spectrophotometry:≥95 % (On dried substance)
- Related substances:(HPLC) Passes test
- Specific optical rotation:+86° to +92° (20°C, 589 nm) (c=1, ethanol) (On dried substance)
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: 23030-2500
- Supplier Number: 23030-0010
- Supplier Number: 23030-0050
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



