Order Entry
United States
Orders LinkContactUsLinkComponent
 

Methyl-2-bromo-5-methoxybenzoate

Toxic
Supplier:   APOLLO SCIENTIFIC
Methyl-2-bromo-5-methoxybenzoate
Supplier:  APOLLO SCIENTIFIC
Certificates
Methyl-2-bromo-5-methoxybenzoate

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Methyl 2-bromo-5-methoxybenzoate, 2-Bromo-5-methoxybenzoic acid methyl ester
  • CAS Number:35450-36-3
  • Formula:C₉H₉BrO₃
  • MDL Number:MFCD00051594
  • Molecular Weight:245.07
  • Boiling Pt:271 °C
  • Density:1.52 g/cm³ (25 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • UN:2810
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Bottle
    10 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $77.17
    $77.17
    Each(1/Itm/Bx)
       $77.17

    Supplier Number: OR23647-10G
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $115.23
    $115.23
    Each(1/Itm/Bx)
       $115.23

    Supplier Number: OR23647-25G
  • Bottle
    100 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $399.18
    $399.18
    Each(1/Itm/Bx)
       $399.18

    Supplier Number: OR23647-100G

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.