1-Iodopropane
:107-08-4
:
:C₃H₇I
:MFCD00001095
:169.99 g/mol
:101…102 °C
:–101 °C
:1.743 g/cm³ (25 °C)
:44 °C
:Refrigerator
:2392
:3,III
:GHS02
:GHS07
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.