ContactUsLinkComponent
Falcon® Feeder tray, 96-Well, Corning
Falcon® Feeder tray, 96-Well, Corning
Catalog # BD-353924
Supplier:  Corning
   Low Manufacturing Impact
undefined
Falcon® Feeder tray, 96-Well, Corning
Catalog # BD-353924
Supplier:  Corning
Supplier Number:  353924
   Low Manufacturing Impact
Specifications
  • Description:
    Feeder Tray and Lid for 96-Well Systems
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental Certification:
    ISO 14001 - Bedford (reg. no. 14.4942)
  • Cat. No.:
    BD-353924
  • Supplier no.:
    353924
Specifications
About this item
B DFalcon® HTS 96-multiwell feeder tray and lid, nontreated, hydrophobic polystyrene surface.
  • Sterile
  • Nonpyrogenic
For use with BD Falcon HTS FluoroBlok* 96-multiwell insert systems (BD351161, BD351162, BD351163, BD351164).