Order Entry
United States
Orders LinkContactUsLinkComponent
2-Bromophenyl methyl sulfone 98%

2-Bromophenyl methyl sulfone 98%

Irritant
Supplier:   APOLLO SCIENTIFIC
2-Bromophenyl methyl sulfone 98%
Supplier:  APOLLO SCIENTIFIC
Certificates
2-Bromophenyl methyl sulfone 98%

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  2-Bromophenyl Methyl Sulfone, 1-Bromo-2-(methylsulfonyl)benzene, 2-Bromophenyl Methyl sulphone, 1-Bromo-2-(methylsulphonyl)benzene
  • CAS Number:33951-33-6
  • Formula:C₇H₇BrO₂S
  • MDL Number:MFCD07368924
  • Molecular Weight:235.1
  • Melting Pt:106…110 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $21.38
    $21.38
    Each(1/Itm/Bx)
       $21.38

    Supplier Number: OR6951-1G
  • Bottle
    10 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $119.35
    $119.35
    Each(1/Itm/Bx)
       $119.35

    Supplier Number: OR6951-10G

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.