Order Entry
United States
Orders LinkContactUsLinkComponent
2-Phenoxyethanol

2-Phenoxyethanol

25 g, Bottle
Catalog # 76584-052
Irritant
Supplier:   APOLLO SCIENTIFIC
Supplier #:  OR4481-25G
2-Phenoxyethanol
Catalog # 76584-052
Supplier:  APOLLO SCIENTIFIC
CAS Number:  122-99-6
2-Phenoxyethanol

25 g, Bottle

CAS Number:  122-99-6
Price
$21.91
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether
  • CAS Number:122-99-6
  • Formula:C₆H₅OCH₂CH₂OH
  • MDL Number:MFCD00002857
  • Molecular Weight:138.17 g/mol
  • Boiling Pt:237…246 °C
  • Melting Pt:41944 °C
  • Density:1.107 g/cm³ (25 °C)
  • Flash Pt:121 °C
  • Storage Temperature:Ambient
  • EINECS:204-589-7
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    25 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.