Acid Fast Fluorescent (AFB) Stain Set Auramine O
76591-140
:
- Pack type:3 bottles/pack
- Description:Acid fast fluorescent (AFB) stain set auramine O
- Size:8 oz.
- Cat. No.:76591-140
- Application:Clinical
Auramine O stain kit.
Contains 8 oz. each one of the following: Auramine O, fluorescent decolorizer, potassium-permanganate
: Ship ground.