Order Entry
United States
ContactUsLinkComponent
Magnesium hydroxide in aqueous solution 80 g/L
Magnesium hydroxide in aqueous solution 80 g/L
Catalog # RCR4485000500A
Supplier:  Ricca Chemical
CAS Number:  
undefined
Magnesium hydroxide in aqueous solution 80 g/L
Catalog # RCR4485000500A
Supplier:  Ricca Chemical
Supplier Number:  R4485000-500A
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Size:
    500 ml
  • Cat. No.:
    RCR4485000500A

Specifications

Specification Test Results

  • Appearance:
    Milky white suspension
  • Assay (vs. EDTA/Eriochrome Black T):
    77 - 83 g/L

Specification Test Results

About this item

 :
 :58.32 g/mol
Boiling Pt:~100 °C
Melting Pt:~0 °C
Storage Temperature:Ambient
CAS Number:1309-42-8
Formula:Mg(OH)₂
MDL Number:MFCD00011104