Panthera TEC EpiPolarizing Trinocular Microscope Packages, Motic
: Motic
The panthera TEC POL models present a high flexibility for the inspection of all kind of polarizing materials, for transparent samples as well as for opaque specimen from technical education environments and industrial quality control.
- Panthera TEC-EpiPolarizing trinocular microscope
- 0.5× C-Mount camera adapter for ¹/₃" and ¹/₂" chip sensors
- Motic analyzes basic 64-bit; USB + dongle (no charge)
- Moticam S6/1080×/BMH4000
The compact Epi illuminator carries a slot for polarizer and analyzer, ready to set up polarization contrast and to reduce internal reflections. An intermediate tube with focusable/centerable bertrand lens is prepared for a conoscopic analyzis of crystals and minerals in transmitted light.
: Contact your avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.
Frequently Bought Together