Linearity FLQ Anti-Mullerian Hormone for Roche Systems, AUDIT MicroControls
Catalog # 76627-722
Supplier: Audit Microcontrols
CAS Number:
Specifications
- Description:Linearity FLQ anti-mullerian hormone for Roche systems
- Size:2 ml
- Cat. No.:76627-722
Specifications
About this item
The Audit® MicroControlsTM Linearity FLQ Anti-Mullerian Hormone for Roche Systems is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analytes: Anti-Mullerian Hormone (AMH).