Linearity FD Procalcitonin for Siemens Atellica, AUDIT MicroControls
Catalog # 76627-720
Supplier: Audit Microcontrols
CAS Number:
Specifications
- Description:Linearity FD Procalcitonin for Siemens Atellica
- Size:5x 1 ml
- Cat. No.:76627-720
Specifications
About this item
The Linearity FD Procalcitonin for Siemens Atellica is assayed quality control material consisting of five levels of bovine based serum albumin. It is intended to simulate human patient serum samples for purpose of determining linearity, calibration verification and verification of reportable range for Procalcitonin.