Corning® BioCoat® Poly-D-Lysine Plates, Corning
Supplier: Corning
Certificates
About this item
Poly-D-Lysine (PDL) and Poly-L-Lysine (PLL) are synthetic compounds that enhance cell adhesion and protein absorption by altering surface charges on the culture substrate. In addition to promoting cell adhesion, poly-lysine surface treatments support neurite outgrowth and improve the survival of many central nervous system (CNS) primary cells in culture. As PDL and PLL are synthetic molecules, they do not stimulate biological activity in the cells cultured on them, and they do not introduce impurities carried by natural polymers.
- Attachment and spreading of a variety of cell types
- Cell differentiation and neurite outgrowth
- Attachment of fastidious transfected cell lines
- Support survival of primary neurons in culture
- Serum-free or reduced-serum culture
- Storage: 4 to 30 °C under dry conditions.
Source: PDL, synthetic (MW 75 to 150 kD); PLL, synthetic (MW 30 to 70 kD).
Specifications
- Catalog No:
- 62406-030
- 62406-026
- 62406-028
- 62406-700
- 62405-749
- 62405-747
- 62406-718
- 62406-024
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental Certification:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
Specifications
Frequently Bought Together