Orders LinkContactUsLinkComponent
PYREX® Separating Funnels, Squibb, Glass, Corning
PYREX® Separating Funnels, Squibb, Glass, Corning
Supplier:   Corning
PYREX® Separating Funnels, Squibb, Glass, Corning
Supplier:  Corning
Certificates
PYREX® Separating Funnels, Squibb, Glass, Corning
PYREX® Separating Funnels, Squibb, Glass, Corning
8 Options Available Below
Product Details & Documents
Borosilicate glass funnels feature an unbreakable PTFE valve and drip tip and have a screw thread closure at the top. Supplied with a PTFE valve assembly and GL screw cap with PTFE-faced silicone liner. No stopper.
Replacement funnels do not include a PTFE valve and drip tip or cap.
Ordering information: For replacement valve bottoms or caps, contact your Avantor representative.
Documents
Product Family Options
Funnels, with bottom valve and cap
Product Information
  • Capacity
    Thread finish
    Availability
    Price
  • 62890-662
    Separatory Funnel with PTFE Valve and Threaded Screw Cap
    Separatory Funnel with PTFE Valve and Threaded Screw Cap
    60 ml
    GL 32
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $193.64
    $193.64
    New Customer Promotional Price
    Case of 1
       $193.64
    Supplier Number: 6305-60
  • 62890-666
    Separatory Funnel with PTFE Valve and Threaded Screw Cap
    Separatory Funnel with PTFE Valve and Threaded Screw Cap
    250 ml
    GL45
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $354.38
    $354.38
    New Customer Promotional Price
    Case of 1
       $354.38
    Supplier Number: 6305-250
  • 62890-668
    Separatory Funnel with PTFE Valve and Threaded Screw Cap
    Separatory Funnel with PTFE Valve and Threaded Screw Cap
    500 ml
    GL45
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $418.30
    $418.30
    New Customer Promotional Price
    Case of 1
       $418.30
    Supplier Number: 6305-500
  • 62890-670
    Separatory Funnel with PTFE Valve and Threaded Screw Cap
    Separatory Funnel with PTFE Valve and Threaded Screw Cap
    1000 ml
    GL45
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $460.31
    $460.31
    New Customer Promotional Price
    Case of 1
       $460.31
    Supplier Number: 6305-1L
  • 62890-672
    Separatory Funnel with PTFE Valve and Threaded Screw Cap
    Separatory Funnel with PTFE Valve and Threaded Screw Cap
    2000 ml
    GL45
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $724.36
    $724.36
    New Customer Promotional Price
    Case of 1
       $724.36
    Supplier Number: 6305-2L
  • 62890-674
    Separatory Funnel with PTFE Valve and Threaded Screw Cap
    Separatory Funnel with PTFE Valve and Threaded Screw Cap
    4000 ml
    GL45
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $1,128.40
    $1,128.40
    New Customer Promotional Price
    Case of 1
       $1,128.40
    Supplier Number: 6305-4L
Funnels, without bottom valve and cap
Product Information
  • Capacity
    Thread finish
    Availability
    Price
  • 62890-678
    Funnel Only
    Funnel Only
    60 ml
    GL 32
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $183.30
    $183.30
    New Customer Promotional Price
    Case of 1
       $183.30
    Supplier Number: 6305-60FO
  • 62890-684
    Funnel Only
    Funnel Only
    500 ml
    GL45
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $303.62
    $303.62
    New Customer Promotional Price
    Case of 1
       $303.62
    Supplier Number: 6305-500FO
  • 62890-688
    Funnel Only
    Funnel Only
    2000 ml
    GL45
    Discontinued
    Supplier Number: -
  • 62890-692
    Funnel Only
    Funnel Only
    6000 ml
    GL45
    Discontinued
    Supplier Number: -
Accessories
Product Information
  • Catalog #
    Size
    Availability
    Price
  • PTFE Valve Bottoms
    PTFE Valve Bottoms
    62890-658
    6 mm
    Log in for availability
    Case of 1 - $195.52
Product Information
  • Catalog #
    Size
    Availability
    Price
  • PTFE Valve Bottoms
    PTFE Valve Bottoms
    62890-660
    8 mm
    Log in for availability
    Case of 1 - $195.52