Corning® Primaria™ Cell Culture Flasks, Corning
Supplier: Corning
Certificates
About this item
PS, tissue culture-treated, sterile, non-pyrogenic
- Nitrogen-containing tissue culture surface chemistry improves attachment, spreading and growth for many primary cells or cell lines
- Vented caps incorporate a 0.2 µm hydrophobic membrane
- Optically clear and no special storage required
- Convenient, peel-open packaging
The complex surface on Corning® Primaria™ products is homogeneous and stable and is used to improve attachment and differentiation of a variety of cell types. The surface chemistry of Corning® Primaria™ products is confirmed by Electron Scanning for Chemical Analysis (ESCA).
Note: Growth area and capacity are nominal