Orders LinkContactUsLinkComponent
Corning® microplates, 384-well, multi-coated
Corning® microplates, 384-well, multi-coated
384, Black with Clear Bottom, Non sterile, Multicoated
Catalog # 75805-282
Corning® microplates, 384-well, multi-coated
Catalog # 75805-282
Supplier:  
Corning® microplates, 384-well, multi-coated
Corning® microplates, 384-well, multi-coated
384, Black with Clear Bottom, Non sterile, Multicoated
Discontinued
Product Details & Documents
Multi-coated microplate, PS, black with clear, flat bottom, with lid. Black wall plates have low background fluorescence, minimal light scatter and reduced crosstalk.
  • Opaque walls to prevent well to well crosstalk
  • Optically clear flat well bottom permits direct microscopic viewing
  • Can be used for both top and bottom reading instruments
  • Individual alphanumeric codes for well identification
Specifications
  • No. of wells:
    384
  • Sterility:
    Non sterile
  • Color:
    Black with Clear Bottom
  • Surface treatment:
    Multicoated