Order Entry
United States
ContactUsLinkComponent
Msz Exonuclease I
Msz Exonuclease I
Catalog # 76595-278
CAS Number:  
undefined
Msz Exonuclease I
Catalog # 76595-278
Supplier Number:  M0527S
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Source:
  • Species:
  • Size:
    1000 units
  • Environmentally Preferable:
  • Enzyme Name:
    Msz Exonuclease I
  • Concentration:
    10000 units/ml
  • Cat. No.:
    76595-278

Specifications

About this item

A DNA specific exonuclease isolated from Methylocaldum szegediense that catalyzes the removal of nucleotides from linear single-stranded DNA in the 3' to 5' direction with optimal activity between 45 and 60 °C. Isolated from a strain of E. coli which carries the cloned pMC45-1 gene from Methylocaldum szegediense.

  • Catalyzes removal of nucleotides from linear single-stranded DNA in 3' to 5' direction
  • Ideal for thermophilic workflows between 45 and 60 °C
  • Can be heat inactivated at 80 °C in 1 minute
  • Supplied with 1 vial of rCutSmart Buffer (10X)