Abbott™ Alere™ hCG Cassette (25 mIU/ml), Healthcare Products
Catalog # 76553-940
Supplier: Healthcare Products
CAS Number:
Specifications
- Description:Alere™ hCG cassette test kit (25 mIU/ml)
- Size:40 tests
- Cat. No.:76553-940
Specifications
About this item
The Alere™ hCG cassette (25 mIU/ml) is a one-step test for the early detection of human chorionic gonadotropin (hCG), a hormone produced during pregnancy.
- Fast - Single-step test provides positive results in as little as 3 minutes
- Easy - CLIA-Waived, easy-to-use test
- Reliable - Highly sensitive assay with built-in control ensures accuracy
The test uses a patented, innovative technology to provide highly sensitive and specific results for pregnancy.