ADAPTER MASTERSENSE AUX/BARE WIRE
MFLX07526-99
- Max. Pressure:100 psi
- Description:Masterflex® universal pressure sensor adaptor cable, bare wire- MasterSense™ pump systems, 6' cable
- Cat. No.:MFLX07526-99
- Includes:6' cable
Adapts a variety of sensors to the MasterSense™ Sensor AUX input.
- Designed for use with 0-20/4-20 mA or 0-5,0-10 V sensors
- Includes 24VDC power out pins for loop powered circuits up to 50 mA
Connection cable allows for integration of variety of sensors into Masterflex® MasterSense™ pump systems. Reference MasterSense™ instruction manual for wiring diagrams and detail.
- Max. Pressure:100 psi
- Description:Masterflex® universal pressure sensor adaptor cable, bare wire- MasterSense™ pump systems, 6' cable
- Environmentally Preferable:
- Cat. No.:MFLX07526-99
- Includes:6' cable