ContactUsLinkComponent
Rotary Pipette Stand for Nichipet, Nichiryo America
Rotary Pipette Stand for Nichipet, Nichiryo America
Catalog # 89078-422
Supplier:  Nichiryo America
Rotary Pipette Stand for Nichipet, Nichiryo America
Catalog # 89078-422
Supplier:  Nichiryo America
Supplier Number:  00-MLT-STD2
Specifications
  • Description:
    Rotary pipette stand for 6 pipettes; Nichipet Premium LT, Nichipet Premium, Nichipet EXII, Nichipet EX PlusII, Nichipet EX II Multi
  • Cat. No.:
    89078-422
  • Supplier no.:
    00-MLT-STD2
Specifications
About this item
Rotary stand made of ABS.
  • Capacity of six single pipettes
For Nichipet Premium LT, Nichipet Premium, Nichipet EXII, Nichipet EX PlusII, Nichipet EX II Multi.