Supelco® Positioning Dollies, Merck
Catalog # EM-695040-1
Supplier: MilliporeSigma
Specifications
- Ø:19 ¼"
- Description:Positioning dolly, for 50 L
- Material:Steel
- Cat. No.:EM-695040-1
- Load capacity:900 lb
- Supplier no.:695040-1
Specifications
About this item
Metal drum dolly for 50 L and smaller drums.