Mini Slide Drying Hotplate
Product Details & Documents
Mopec’s mini slide drying hotplate is designed to quickly and efficiently remove wrinkles from tissue sections while taking up minimal bench-top space. The rapid heating system ensures the hotplate is ready to use in less time.
Dual Voltage: 110 and 230 V power supplies
Designed to take up minimal bench-top space
- Rapid heating system ensures system is ready to use in less time
- Anti-microbial coating to help prevent bacteria and virus growth
Sold individually.
Specifications
- Description:Mini slide drying hotplate
- Electrical:110 or 230 V